Transfection reagent, in vivo-jetRNA
Supplier: Polyplus-transfection
101000021.EA 1760 EUR
PPLU101000021
Transfection reagent, in vivo-jetRNA
in vivo-jetRNA® is the reagent of choice to deliver mRNA in vivo. Being a non viral delivery reagent, in vivo-jetRNA® allows safe administration in animals via different administration routes.
  • Adaptable: Suited for any injection route to target any organ
  • Safe: Maintains healthy animals and organs
  • Time saving: Ready to use reagent with an easy protocols
  • Tailored: Customised protocols from in vivo delivery experts
in vivo-jetRNA® is a transfection reagent specifically developed to deliver mRNA in vivo. in vivo-jetRNA® can be used to target multiple organs, by using systemic or local injection routes, in various animal models. mRNA delivery using in vivo-jetRNA® is user-friendly, with a simple 2-step protocol and can be used for vaccination purposes, anti-cancer studies, genome editing using CRISPR/Cas9 method or protein replacement.
Certifications: Since 2002, Polyplus-transfection® is ISO 9001 certified, and since then this has been successfully renewed every three years.