Cleanroom gloves, latex, BioClean Legacy™
Supplier: Nitritex
BioClean Legacy™
112-3304EA 306 EUR
112-3304 112-3303 112-3308 112-3307 112-3306 112-3305
Cleanroom gloves, latex, BioClean Legacy™
Gloves
These ambidextrous cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments. Suitable for double-donning.
  • Protein level less than 50 µg/g
  • Textured finish for improved grip
  • Latex provides exceptional flexibility and comfort
  • Beaded cuff provides easier donning and stability on the arm
  • Each size carries a unique barcode on the carton (ITF-14), which allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
Colour: Natural
EN 455, EN 374, Cat. III; AQL 1.5
Certifications: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Packaging: Packaging: 100 pieces per sealed inner PE bag; one sealed inner PE bag per sealed outer PE bag; 10 outer PE bags per carton liner; one carton liner per carton (1000 pieces).
Packaging: 100 pieces per sealed inner PE bag; one sealed inner PE bag per sealed outer PE bag; 10 outer PE bags per carton liner; one carton liner per carton (1000 pieces).