Electrical resistant glove liners, A0150 / A0160 / A0170
Supplier: Best
Conductive Fit
112-4217EA 84.1 EUR
112-4217 112-4216 112-4215 112-4214 112-4213 112-4212 112-4211 112-4210 112-4218
Electrical resistant glove liners, A0150 / A0160 / A0170
Gloves
These low linting nylon/acrylic knitted gloves have anti-static properties. The conductive Thunderon® fibre covers the important area of the fingers, providing static discharge by corona effect. The gloves are available without coating, with PU coating on fingertips only, and with coating on fingertips and palm.
  • Surface resistivity of 106 to 107 Ω
  • Optimal dexterity and tactile feel
  • Seamless knit: No skin irritation
  • Elasticated cuffs: Great freedom of movement for the wrist
  • PU coating protects extremities from oils and micro cuts and prevents fingerprints on handled objects
Ideal for use when working with electronic products, in manufacturing industries and in automotive-related industries.
Colour: Light grey/white
A0150/A0160: EN 388 (0130); Cat. II
A0170: EN 388 (3121); Cat. II
Certifications: A0150: EN 388:2016 (0130X); Cat. II
A0160: EN 388:2016 (0130X), EN 16350:2014; Cat. II
A0170: EN 388:2016 (3121X), EN 16350:2014; Cat. II