ESD shoe covers, BioClean ESD™
Supplier: Nitritex
BioClean ESD™
113-0815EA 600 EUR
113-0815 113-0816 113-0817
ESD shoe covers, BioClean ESD™
Disposable Garments
The anti-static ESD disposable overshoes with conductive tape are manufactured from non woven spunbonded polypropylene.
  • Superior ESD performance
  • Fitted with a polyester conductive ribbon with a conductive carbon filament
  • Non marking ribbon
  • Low levels of particulate shedding and double bagged for cleanroom use
Suitable for use in ISO class 5 cleanrooms.
Length: 400 or 455 mm