Cleanroom face masks with visors, BioClean Clearview™, PE/PP
Supplier: Ansell-Nitritex
BioClean Clearview™
NITRVFM210-T-SLOTEA 428 EUR
NITRVFM210-T-SLOT 113-2266
Cleanroom face masks with visors, BioClean Clearview™, PE/PP
Masks Cleanroom Masks
Face masks made from 3 ply, PP/PE, ultrasonically bonded fabric with integrated anti-fog visors. The outer facing consists of hydrophobic non woven PE, the filter layer of meltblown PE and the inner facing of hydrophilic PP/PE (Non woven).
  • No latex proteins
  • Particle filtration efficiency (PFE): >97%, bacterial filtration efficiency (BFE): >96,5%
  • Barcodes appear on the outer carton to allow automatic data entry into stock control systems
  • With ties or ear loops
Suitable for use in class 10, ISO 4, M2.5 environments.
Thickness (visor): 0,18 mm
Certifications: Manufactured in a facility holding ISO 9001:2008. Processed in an NEBB certified ISO class 4 cleanroom environment. Sampling and inspection in accordance with ISO 2859-3:1993.