VWR®, Cleanroom masks
Supplier: VWR
VWR®
113-9162EA 37.6 EUR
113-9162 113-9163
VWR®, Cleanroom masks
Masks Cleanroom Masks
Cleanroom masks made from non woven fabric featuring ties.
  • Latex-free
  • High bacterial filtration efficiency
  • Sterile version with the sterility assurance level of 10⁻⁶
  • For industrial and cleanroom use only, do not use for protection against COVID-19
  • This product is not a medical device for medical or clinical use within the meaning of Regulation (EU) 2017/745 / Directive 93/42/EEC
  • This product is not personal protective equipment (PPE) within the meaning of Regulation (EU) 2016/425
Density: 20 g/m² (front layer); 25 g/m² (back layer); 28 g/m² (centre).
Certifications: Meets IEST level 2 (2440 counts/ft³ for ≥0,5 µ).
Packaging: 50 masks per pack, 10 packs per case.