VWR®, Cleanroom Wipes, 100% Polyester, Double Ply
Supplier: VWR
VWR®
115-2446EA 1060 EUR
115-2446 115-2470 114-9323
VWR®, Cleanroom Wipes, 100% Polyester, Double Ply
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These wipes are made from 100% continuous filament, double-ply, ultrasonically-sealed polyester to provide better absorbency.
  • Free from silicone, amides and DOP contamination
  • Low particle and fibre levels
  • Premium border sealed; very soft edges, no linting
  • Highly absorbent
  • Laundered and packed in ISO 4 cleanroom environment
Ideal for wiping the interior of process tools and other equipment, as well as for spill control and general wiping in ISO 4 and above environments. Suitable for use in pharmaceutical production, microelectronic facilities, and other high-tech industries.