Biological safety cabinet, BioVanguard B, for cytotoxic (high risk) applications (cytotoxic medicines)

Supplier: CleanAir by Baker
135-1871EA 17210 EUR
135-1871 135-1865 135-1867 135-1869
Biological safety cabinet, BioVanguard B, for cytotoxic (high risk) applications (cytotoxic medicines)
Fume Hoods and Enclosures Biological and Microbiological Safety Cabinets
BioVanguard Greenline B is a class II cabinet range which offers simplicity, robustness and high reliability ensuring the highest level of protection for operator, product and the environment, minimising hazards inherent to working with agents assigned to biosafety levels 1, 2 and 3. BioVanguard B is designed for high risk microbiological and high toxic applications, such as the production of cytotoxic medicines.

  • Easy to install, easy to operate, easy to clean, easy to decontaminate with low maintenance
  • Microprocessor control with automatic filter clogging compensation
  • User-friendly LCD control panel, positioned at an angle, showing the operator all relevant functions and alarm (visual and audible) at a glance
  • Ergonomic epoxy coated aluminium arm rest contributes to comfortable working position of the operator
  • Two independent blowers and anemometers to control chamber air flow and inlet/exhaust flow
  • Exhaust and chamber HEPA filters H-14 of efficiency 99,995 % MPPS (99,999 % DOP)
  • RS232 and RS485 connection

BioVanguard B is equipped with:
- Epoxy coated exterior
- Stainless steel interior
- Stainless steel worktop
- Pre-filter and drip tray
- 2 × electrical sockets

BioVanguard B is equipped with additional V-shaped HEPA filters underneath the work surface, which filter the inflow air and keep the internal construction of the cabinet free of contamination therefore ensuring the utmost safety for high risk applications.

The stainless steel work-top with V-shaped air slits provides superior safety by preventing the inflow from blocking and the contamination of operator’s sleeves. The front window is made of laminated safety glass and can be completely hinged, opening the entire work area for easy cleaning and/or (un)loading large items.

Available with manual or electric windows.

Certifications: BioVanguard Series is designed and manufactured according to international biosafety standards EN12469: Class II, NSF49:
Class II Type A2, JIS K 3800: Class II Type A2 and PIC’s. The working area is classified as ISO 14644: Class 5 and GMP
Annex 1: Class A. BioVanguard B is also designed and manufactured according to DIN 12980: Class II.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR