Biological safety cabinets, Herasafe™ 2030i
Supplier: Thermo Scientific
HERAsafe®
135-2661EA 21770 EUR
135-2661 135-2660 135-2663 135-2662 135-2665 135-2664 135-2667 135-2666
Biological safety cabinets, Herasafe™ 2030i
Fume Hoods and Enclosures Biological and Microbiological Safety Cabinets
HeraSafe 2030i biological safety cabinets have a large interior workspace made of 316 non-corrosive stainless steel for incorporation of instruments and process optimisation devices, including robotic arms.
  • Controlled passcode access
  • Interior LED lights
  • Event and alert messaging logs
  • Standard smart ports
  • Padded armrests
  • Self-adjusting germicidal UV light intensity
Equipped with smart self-monitoring safety features, intuitive graphic touch screen interface and built-in connectivity to preserve important performance data. Electronic sash with fully hinged opening enables easy access for supplies/equipment and simplifies easy cleaning and disinfection.
Digital airflow verification system validates protection by monitoring airflow velocities with independent pressure sensors which detect and alert to any changes across the exhaust and downflow plenums.
Certifications: EN 12469.