Herasafe™ 2030i Biological Safety Cabinets
Supplier: Thermo Fisher Scientific
HERAsafe®
135-2661EA 24130 EUR
135-2661 135-2660 135-2663 135-2662 135-2665 135-2664 135-2667 135-2666
Herasafe™ 2030i Biological Safety Cabinets
Fume Hoods and Enclosures Biological Safety Cabinets
HeraSafe 2030i biological safety cabinets have a large interior workspace made of 316 non-corrosive stainless steel for incorporation of instruments and process optimisation devices, including robotic arms.
  • Controlled passcode access
  • Interior LED lights
  • Event and alert messaging logs
  • Standard smart ports
  • Padded armrests
  • Self-adjusting germicidal UV light intensity
Equipped with smart self-monitoring safety features, intuitive graphic touch screen interface and built-in connectivity to preserve important performance data. Electronic sash with fully hinged opening enables easy access for supplies/equipment and simplifies easy cleaning and disinfection.
Digital airflow verification system validates protection by monitoring airflow velocities with independent pressure sensors which detect and alert to any changes across the exhaust and downflow plenums.
Self-adjusting germicidal UV light intensity - Our unique cross beam UV system has exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use.
Bright work area with glass side walls and LED lights. SmartPort convenience easily pass tubing and wiring through sidewall to accommodate many applications needs.
Alarms, alerts, system self-checks and performance status are provided on a full-color touchscreen. Touchscreen and connectivity (available in select countries) - Built-in WiFi system is ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools and more. Digital Airflow Verification (DAVe) alerts, system self-checks and performance status are provided on the full-color touchscreen.
Certifications: EN 12469. The 1,2 m model has been certified to EN 12469 with the NF mark by LNE as well.