Colorimetrics tubes for gas measurements
Supplier: Gastec
Airtec
GAST18ELEA 31.7 EUR
GAST18EL 151-0061 GAST211LL 118-0468 151-0056 151-0055 118-0467 151-0058 118-0466 118-0465 151-0057 151-0059 118-0469 118-0460 763-0290 118-0464 118-0463 118-0462 118-0461 GAST17LL GAST45S 151-0052 151-0051 151-0054 151-0053 118-0358 151-0045 118-0357 151-0044 151-0047 151-0049 151-0048 118-0359 118-0471 118-0470 118-0475 118-0474 118-0472 151-0041 GAST172 GAST171 151-0040 151-0043 151-0042 151-0036 118-0047 151-0038 151-0037 151-0039 GAST131L 151-0030 GAST71H 151-0031 118-0457 118-0456 118-0455 151-0025 151-0024 118-0454 151-0027 151-0026 151-0029 118-0459 118-0458 151-0028 118-0052 GAST172L 118-0051 118-0453 118-0452 118-0451 118-0450 709-0060 151-0020 151-0012 151-0011 151-0014 151-0013 GAST40 151-0016 151-0015 151-0018 151-0017 151-0019 151-0010 GAST211H 151-0001 151-0003 151-0002 151-0005 151-0004 151-0007 151-0006 GAST211M 151-0009 151-0008 GAST131LA GAST131LB GAST131 118-0369 118-0368 118-0367 118-0366 GAST111L 118-0361 118-0360 118-0365 118-0486 118-0364 118-0363 118-0362 763-0286 763-0285 118-0378 763-0288 118-0377 763-0287 763-0289 118-0372 118-0370 118-0376 118-0375 763-0284 118-0374 763-0283 118-0373
Colorimetrics tubes for gas measurements
Gas Tubes
Crystal clear gas tubes with distinct colour changes.
  • Easy to read calibration scale
  • Specially adapted for accurate direct-reading measurements
These tubes have to be used with a pump.
Certifications: EN 1231.
Other tubes on request.
Ordering information: Other tubes are available on request.