Chromatography Tubing, PTFE
Supplier: VWR
16-PTFE-30-5MEA 116 EUR
16-PTFE-30-5M 16-PTFE-30-10M 08-PTFE-62-3M 08-PTFE-62-10M 08-PTFE-62-5M 16-PTFE-07-10M 16-PTFE-07-5M 16-PTFE-20-5M 16-PTFE-20-10M 16-PTFE-20-3M 16-PTFE-07-3M 16-PTFE-30-3M
Chromatography Tubing, PTFE
Tubing Chromatography Tubing
PTFE
  • Flexible and economical
  • Excellent solvent resistance
Inert to all chemicals used in HPLC but suitable for low-pressure applications only. The most common tubing used to connect the solvent reservoir to the pump.