ETFE tubing
Supplier: Cytiva
18-1120-96EA 61 EUR
18-1120-96 18-1121-36
ETFE tubing
Tubing Transfer Tubing
Ethylenetetrafluorethylene tubing (ETFE) is designed for use with AKTA systems.
White/clear tubing for easy identification.