XK and K 9 series O-rings
Supplier: Cytiva
19-0681-01EA 23.52 EUR
19-0681-01 19-0685-01
XK and K 9 series O-rings
Seals Sealing Rings
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules with the following few exceptions - chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used.