Hot Start Taq PCR Master Mix, 2X
Supplier: VWR Life Science
1B1408-100RXNEA 0 EUR
1B1408-100RXN
Hot Start Taq PCR Master Mix, 2X
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hot Start Taq Master Mix, 2X is a ready to use reaction cocktail that contains all required components, except primers and template DNA, for routine PCR amplification of DNA fragments up to 4 kb. The included Taq DNA polymerase is inactivated with proprietary monoclonal antibodies. Incubation at ≥94 °C during PCR thermal cycling irreversibly denatures the antibody and releases full Taq DNA polymerase activity.