Dropping funnels, cylindrical, without pressure balance
Supplier: VWR, part of Avantor
VWR®
201-1240EA 14 EUR
201-1240 201-1238 201-1239 201-1241 201-1242 201-1243 201-1244
Dropping funnels, cylindrical, without pressure balance
Funnels Dropping Funnels
Borosilicate 3.3 glass.
  • Ground glass cone for flasks and assemblies
  • PTFE key stopcock and PP stopper
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
PTFE key stopcock for non stick, grease-free performance.
Certifications: ISO 4800