Dropping funnels, cylincrical, with pressure balance
Supplier: VWR, part of Avantor
VWR®
201-1250EA 89.7 EUR
201-1250 201-1249 201-1245 201-1246 201-1247 201-1248
Dropping funnels, cylincrical, with pressure balance
Funnels Dropping Funnels
Borosilicate 3.3 glass.
  • Standard tapered joints top and bottom, drip stem and pressure-equalising arm
  • PTFE key stopcock and plastic stopper
  • Very good chemical resistance
  • Very good temperature resistance
Drip stem does not extend beyond joint, thereby eliminating a possible cause of breakage. PTFE key for grease-free performance.