Sorbent tubes for gas and vapor sampling, ORBO™
Supplier: Merck
ORBO™
20264-UEA 159 EUR
20264-U 20267-U 20050-U 20030-U 20362. 54020-U 54081-U 20358. SUPL20211 SUPL20051 SUPL20351 SUPL20053 20254-U 20256-U 20255-U 20257-U 20832-U 20081-U 23446-U 20355. SUPL20265 SUPL20044 20349. SUPL20360 SUPL20063 SUPL20263 SUPL20262
Sorbent tubes for gas and vapor sampling, ORBO™
Gas Tubes
ORBO™ sorbent tubes contain two beds of the same selective adsorbent seperated by glass wool or foam, for gas and vapor sampling.
  • Wide range of high quality adsorbents
  • Consistent adsorbent mesh sizes
  • Coated and specially-treated materials
  • Reproducible recoveries and low standard deviation
  • Consistent bed weights from tube to tube to ensure capacity
The dual-layer or two-bed construction of the tube allows for any sample breakthough to be captured in the smaller back-up bed.