Microcentrifuge tubes, 0,70 ml, with snap cap
Supplier: Kartell
Kartell
211-5811EA 40.7 EUR
211-5811 211-5812 211-5813 211-5814
Microcentrifuge tubes, 0,70 ml, with snap cap
Tubes Microcentrifuge Tubes
Microcentrifuge tubes with caps, made of HDPE, for Cobas Bio® and Cobas Mira® automatic analyzers.
  • Good chemical resistance
  • Good temperature resistance
Certifications: CE and IVD