Safety waste systems, SP Bel-Art
Supplier: Bel-Art
212-0731EA 303 EUR
212-0731
Safety waste systems, SP Bel-Art
Bottles Safety Waste Bottles
HDPE, translucent, graduated.
  • Translucent for observing content level
  • Includes screw-top to seal container for safe transit
  • Flared and angled funnel for easy filling
A system for safe handling and temporary storage of spent solvents or other liquid waste. Features a screw-on funnel top with snap-on cap seals to retain fumes.