Beakers, low form
Supplier: Kartell
Kartell
213-0191EA 37.1 EUR
213-0191 213-0190 213-0185 213-0184 213-0183 213-0189 213-0188 213-0187 213-0186
Beakers, low form
Beakers
PMP (TPX), crystal clear.
Ideal for general laboratory use. With spout and easy to read embossed graduations.
  • Good chemical resistance
  • Heat resistant up to +150 °C (up to +170 °C for short periods), autoclavable
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read embossed graduations.
  • Good chemical resistance
  • Heat resistant up to +150 °C (up to +170 °C for short periods), autoclavable
  • Smooth surfaces for easy, fast cleaning
ISO 7056
Certifications: ISO 7056