Beakers, low form
Supplier: Kartell
Kartell
213-0192EA 3.8 EUR
213-0192 213-0196 213-0195 213-0194 213-0193 213-0200 213-0199 213-0198 213-0197
Beakers, low form
Beakers
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read moulded graduations.
Certifications: ISO 7056