Beakers, Philips, DURAN®
Supplier: DWK Life Sciences
213-0254EA 28.34 EUR
213-0254 213-0215 213-0214
Beakers, Philips, DURAN®
Beakers
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Thick-walled, with printed graduations and labelling area.