VWR®, Beakers, Low Form, PP
Supplier: VWR
213-0490EA 6.5 EUR
213-0490 213-0491 213-0492 213-0493 213-0489 213-0511 213-0494 213-0495 213-0496
VWR®, Beakers, Low Form, PP
Beakers
PP, translucent.
  • Printed graduations
  • Can be stacked when empty to save space while storing
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements
Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.