Beakers, low form, DURAN®
Supplier: DWK Life Sciences
213-1107EA 10 EUR
213-1107 213-1108 213-1109 213-1114 213-1103 213-1115 213-1104 213-1105 213-1106 213-1110 213-1111 213-1112 213-1101 213-1113 213-1102
Beakers, low form, DURAN®
Beakers
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area (213-1101, 213-1102 not graduated and without Retrace Code).
With printed Retrace Code (batch identification).
ISO 3819 (except 213-1115).