Beakers, low form
Supplier: Kartell
Kartell
213-1636EA 13.7 EUR
213-1636 213-1631 213-1632 213-1643 213-1633 213-1644 213-1645 213-1634 213-1641
Beakers, low form
Beakers
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use at +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
Certifications: ISO 7056