Beakers, low form
Supplier: Kartell
Kartell
213-1914EA 12 EUR
213-1914 213-1915 213-1916 213-1917 213-1911 213-1912 213-1913 213-1918 213-1919
Beakers, low form
Beakers
PMP (TPX), crystal clear.
  • Good chemical resistance
  • Heat resistant up to +150 °C (up to +170 °C for short periods), autoclavable
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
  • Good chemical resistance
  • Heat resistant up to +150 °C (up to +170 °C for short periods), autoclavable
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
Certifications: ISO 7056