Measuring beakers with handle
Supplier: Kartell
Kartell
213-3430EA 14.6 EUR
213-3430 213-3440 213-1625 213-3425 213-3420
Measuring beakers with handle
Beakers
PP, translucent.
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning