Bottles, rectangular, narrow neck with screw caps
Supplier: Gosselin
215-0699EA 368 EUR
215-0699 215-0701 215-0700 215-0702
Bottles, rectangular, narrow neck with screw caps
Bottles Plastic Bottles
HDPE, translucent, graduated with red tamper evident screw caps.
  • Rectangular cross section bottles save storage space
  • Graduated bottles are supplied with caps but are unassembled
  • Sterile bottles are irradiated and are supplied capped
These bottles are graduated and sterile available. Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.