Bottles, wide neck, with screw cap
Supplier: Gosselin
215-0754EA 247 EUR
215-0754 215-0743 215-0761 215-3962 GOSSLR100-13 GOSSLR100-07 215-3972 215-0752 215-3239 215-0741 215-3238 GOSSLR500-07 215-0758 215-0770 215-0764 215-0748 215-3968 215-0745 215-0765 GOSSLR150A-06 215-3969 215-3977 215-0756 215-0766 215-0760 215-3965 215-0755 215-3974 215-3971 215-0762 215-0749 215-0744 GOSSLR100-08 GOSSLR500-15 215-3290 215-3963 215-0768 215-0759 215-0767 215-2184 215-1295 GOSSLR150A-08 215-0769 215-3240 215-0753 215-2183 215-0751 215-0740 GOSSLR100-10 215-3973 215-0747 215-3970 215-0742 215-0771
Bottles, wide neck, with screw cap
Bottles Plastic Bottles
HDPE, translucent with screw caps.
  • Bottles are supplied capped aseptic or ionised
  • Excellent shock resistance
  • Wide neck, easy to fill
A range of round wide mouth bottles. Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.