Sample containers, with snap-on caps
Supplier: Corning Life Sciences
Gosselin
216-0417EA 233 EUR
216-0417 216-2082 216-4277 216-2081 216-2083 216-4278 215-0794 215-0793 GOSSPC1000-09
Sample containers, with snap-on caps
Sample Containers General Purpose Containers
PP, natural or opaque white, with or without snap caps.
  • Excellent shock resistance
  • Graduated or non graduated
  • Tight seal
Ideal for sampling, storage and transport of liquid and solid samples.
Ordering information: Available with or without snap cap (assembled or non-assembled).
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904