Bottles, narrow neck, with standard ground stoppers, DURAN®
Supplier: DWK Life Sciences
215-1735EA 21.47 EUR
215-1735 215-1016 215-1484 215-1485 215-1482 215-1020 215-1021 215-1483 215-1282
Bottles, narrow neck, with standard ground stoppers, DURAN®
Bottles Glass Bottles
Soda-lime glass, clear or brown, with standard ground joint and glass stopper.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and tough
Available in crystal clear glass or brown tinted to protect light-sensitive substances.
Certifications: DIN 12036, ISO 4796