Bottles, narrow neck, square, HDPE, with screw cap
Supplier: Gosselin
215-0655EA 275 EUR
215-0655 215-0643 215-0665 215-0657 215-0662 215-0650 215-3897 215-0661 215-3896 215-1283 215-0647 215-0658
Bottles, narrow neck, square, HDPE, with screw cap
Bottles Plastic Bottles
HDPE translucent, with LDPE screw cap.
  • Square cross section bottles saves on storage space
  • Bottles are available capped (assembled) or uncapped (unassembled)
A range of square, narrow mouth bottles. Ideal for use in the laboratory, production plant or in the field for storage of liquids, solids and powders.