Containers, wide mouth, with screw cap
Supplier: Gosselin
215-1309EA 271 EUR
215-1309 215-1311 215-1312 215-1323
Containers, wide mouth, with screw cap
Sample Containers General Purpose Containers
PP, opaque white, with screw cap.
  • Excellent shock resistance
  • Very good chemical resistance
Ideal for sampling, storage and transport of solid samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904