Bottles, wide neck, with standard ground joint stoppers, DURAN®
Supplier: DWK Life Sciences
215-1318EA 172 EUR
215-1318 215-1319 215-1349 215-1316 215-1317 215-1314 215-1347 215-1348 215-1315 215-1345 215-1313 215-1346 215-1343 215-1321 215-1344 215-1320 215-1350 215-1351
Bottles, wide neck, with standard ground joint stoppers, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
With printed retrace code (batch identification).
Certifications: ISO 4796-2.