Laboratory bottles, narrow neck, with screw cap
Supplier: VWR, part of Avantor
VWR®
215-0059EA 549 EUR
215-0059 215-1592 215-1596 215-0057 215-0058 215-3261 215-1594 215-1595 VWRI215-1596_U 215-1593
Laboratory bottles, narrow neck, with screw cap
Bottles Glass Bottles
Borosilicate 3.3 glass, with PP screw cap and pouring ring.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Economical alternative for a wide range of laboratory applications. Round, with graduations, DIN GL 32 or GL 45 thread.
Certifications: ISO 4796