Bottles, wide neck, HDPE, rectangular or square, with screw cap, VWR®
Supplier: VWR, part of Avantor
VWR®
215-1909EA 45.2 EUR
215-1909 215-1907 215-1908 215-1905 215-1906 215-1904
Bottles, wide neck, HDPE, rectangular or square, with screw cap, VWR®
Bottles Plastic Bottles
HDPE.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof
For collecting, storing and shipping samples, or storing liquids and dry substances. Space saving design with rectangular or square base. 2 and 4 litre bottles have integrated handle design.