Bottles, narrow neck, with screw cap, VWR®
Supplier: VWR, part of Avantor
VWR®
215-2337EA 320 EUR
VWRI215-2328B 215-2337 215-2326 215-2338 215-2327 215-2330 215-3275 VWRI215-2325B 215-2328 215-2325 215-2329 VWRI215-3275B
Bottles, narrow neck, with screw cap, VWR®
Bottles Glass Bottles
Amber borosilicate 3.3 glass with PP screw cap and pouring ring.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of laboratory applications. Round, with graduations, DIN GL 45 thread, pouring ring, and blue PP screw cap. Amber coloured to protect light-sensitive materials.
Purely external application of the colour ensures unchanged glass properties on the inner surface of the bottles.
Certifications: ISO 4796