Wash bottles, narrow or wide neck, standard Line, VWR®
Supplier: VWR, part of Avantor
VWR®
215-2506EA 33.1 EUR
215-2506 215-2533 215-2529 215-2517 215-2514 215-2527 215-2497 215-2500 215-2519 215-2496 215-2499 215-2498
Wash bottles, narrow or wide neck, standard Line, VWR®
Bottles Wash Bottles
LDPE, translucent, with coloured, PP spray cap.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Colour coded caps for quick identification
Economical, soft and very easy to squeeze. For use with general solutions such as water, soap, dilute acids and alkalis. Dispensing tube with pointed end made of PP to optimise backflow of the medium.