VWR® Safety Wash Bottles, Wide Neck, Vented

Supplier: VWR

VWR®
215-2548EA 46.9 EUR
215-2548 215-2538 215-2535 215-2536 215-2540 215-2539
VWR® Safety Wash Bottles, Wide Neck, Vented
Bottles Wash Bottles
LDPE, translucent, with PP vented screw cap.

  • Chemical name in English, French, German and Spanish
  • Chemical formula, CAS number
  • Hazard pictogram, signal word, hazard statements H; precaution statements P

Safety imprint in accordance with regulation (EC) No. 1272/2008 (GHS). Dispensing tube with jet tip made of PP to optimise backflow of the medium that can be cut to length.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR