Spray bottles with pump vaporiser, LaboPlast®
Supplier: Bürkle
LaboPlast®
215-3091EA 2.4 EUR
215-3091 215-3090 215-3092 215-3093
Spray bottles with pump vaporiser, LaboPlast®
Bottles Spray Bottles
Bottle made of HDPE, spray pump made of PP and stainless steel.
  • Good chemical resistance
  • Good temperature resistance
Spray bottles with pump vapouriser and safety cap for the pump. Liquids can be finely vapourised at the push of a button in an environmentally compatible way. The transparent bottles make identification of contents easy. The protective cover prevents unintentional pressing of the pump and acts as a dust cover.