Bottles, wide neck, square, with screw cap, 1000 to 2700 ml capacity
Supplier: Gosselin
GOSSLC1000-03EA 125 EUR
GOSSLC1000-03 215-0718 215-0719 215-3929 GOSSLC2000-17 215-3956 215-0720 215-0722 215-3954 215-3952 215-3951 215-3950 215-3255 215-3254 215-3251 215-1291 215-1292 215-3271 GOSSLC2500A-09 GOSSLC2000-08 212-5185 215-0708 GOSSLC2000-04 GOSSLC2000-07 215-0710 215-3248
Bottles, wide neck, square, with screw cap, 1000 to 2700 ml capacity
Bottles Plastic Bottles
Space saving bottles made of HDPE, translucent with HDPE or LDPE screw cap.
  • Good chemical resistance, good temperature resistance
  • Available sterile or packed under aseptic conditions
  • Excellent shock resistance
Ideal for liquid, solid and powder storage. Good level of tightness according to the closure system used and application.