Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Supplier: Tournaire
TOUR5243S1EA 10.7 EUR
TOUR5243S1 215-3259 215-2949 215-2945
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Bottles Metal Bottles
Pure aluminium.
  • Unbreakable, odorless
  • Very good temperature resistance
  • Leakproof with tamper-evident screw caps (must be odered separately)
  • UN-approved
Suitable for storage and transport of highly aggessive liquids.
Suitable for storage and transport of highly aggessive liquids.