Bottles, wide neck, square, with screw cap, Corning® Gosselin™
Supplier: Corning
Corning®
GOSSLC1000-03EA 125 EUR
GOSSLC1000-03 GOSSLC500-23 215-0718 215-0717 215-0719 215-0714 215-0716 GOSSLC2000-17 215-3956 215-0720 215-0722 215-3954 215-3952 215-3951 215-3950 215-3274 215-1293 215-3273 215-1291 215-3272 215-1292 215-3271 GOSSLC2000-08 212-5185 GOSSLC2000-07 215-0732 215-0734 215-3249 215-3248 215-0730 215-3961 215-3960 GOSSLC500-05 GOSSLC250-06 GOSSLC250-03 GOSSLC500-03 GOSSLC500-04 215-3929 GOSSFC520-10 215-0735 GOSSFC520-04 GOSSFC520-08 GOSSFC520-06 215-3258 215-3257 215-3256 215-3255 215-3254 215-3253 GOSSLC150A-05 215-3252 215-3251 GOSSLC150A-03 GOSSLC2500A-09 GOSSLC520-02 GOSSLC500-10 GOSSFC500-12 GOSSLC500-11 215-0708 215-0710
Bottles, wide neck, square, with screw cap, Corning® Gosselin™
Bottles Plastic Bottles
Space saving bottles made of HDPE, translucent with HDPE or LDPE screw cap.
  • Good chemical resistance, good temperature resistance
  • Sterile oder packed under aseptic conditions
  • Excellent shock resistance
Ideal for liquid, solid and powder storage. Good level of tightness according to the closure system used and application.