Laboratory bottles, wide neck
Supplier: VWR, part of Avantor
VWR®
215-3773EA 239 EUR
215-3773 215-3747 215-3743 215-3746 215-3749 215-3748 215-3745 215-3744
Laboratory bottles, wide neck
Bottles Glass Bottles
Bottles constructed of borosilicate 3.3 glass, in clear or amber colour.
  • Excellent chemical resistance, high temperature resistance
  • 80 mm opening allows easy cleaning
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The large opening can even accommodate large volume funnels, giving easy access to scoops or spatulas.
Amber to protect light-sensitive materials.
Certifications: In compliance with DIN ISO 4796-1 standards.
Delivery information: Supplied with screw cap and pouring ring.
Packaging: The bottles are packed in shrinked-wrapped modules.