Dropping bottles, VWR®
Supplier: VWR, part of Avantor
VWR®
215-3820EA 21.9 EUR
215-3820 215-3823 215-3816 215-3821 215-3818 215-3824 215-3817 215-3828 215-3826 215-3825 215-3822 215-3827 215-3819
Dropping bottles, VWR®
Bottles Dropping Bottles
Soda-lime glass, amber.
  • Good chemical and physical properties
  • Delivered in practical safe trays
  • Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.