Bottles, narrow neck, aluminium, with thread, CHEM 42™
Supplier: Tournaire
TOUR1447S1EA 8.3 EUR
TOUR1447S1 TOUR6440S1 215-3857 215-3856 215-3855
Bottles, narrow neck, aluminium, with thread, CHEM 42™
Bottles Metal Bottles
Pure aluminium (99,5%).
  • High mechanical performance
  • Perfect chemical
  • Totally recyclable
Suitable for storage and transport of highly aggessive liquids.
Suitable for storage and transport of highly aggessive liquids.