Bottles, wide neck, square or rectangular, with screw caps, VWR®
Supplier: VWR, part of Avantor
215-4253EA 140 EUR
215-4253 215-4258 215-4249 215-4361 215-4252 215-4251 215-4255 215-4259 215-4254 215-4248 215-4257 215-4363 215-4256 215-4362 215-4250 215-4247
Bottles, wide neck, square or rectangular, with screw caps, VWR®
Bottles Plastic Bottles
Bottles are made with High Density Polyethylene (HDPE) and are available in a natural or amber finish (2,7 L only). Choose from either square or rectangular bottles to meet specific storage requirements.
  • Sterile 10⁻³
  • Wide mouth design for easy filling
  • Graduation marks for reference during sample collection
  • Packed in smaller packs for better handling and to reduce contamination
  • CE IVD Directive 98/79/EC
Ideal for the storage of liquids, solids, and powders. These bottles are generally used for large volume urine sample collection. Bottles supplied with cap and EPE liner provide an excellent seal between bottle and cap. Natural (translucent) HDPE ensures good visibility of sample content levels. 2,5 and 2,7 L bottles are supplied with a carrying handle.
Certifications: Material resin in compliance with US FDA 21 CFR requirement.