VWR® Safety Wash Bottles
Supplier: VWR
VWR®
215-4309EA 39.1 EUR
215-4309 215-4308 215-4307 215-4306 215-4305 215-4304 215-4303 215-4302 215-4301 215-4323 215-4300 215-4322 215-4321 215-4320 215-4319 215-4318 215-4317 215-4316 215-4315 215-4314 215-4313 215-4312 215-4311 215-4299 215-4310 215-4298 215-4297 215-4296 215-4295 215-4294
VWR® Safety Wash Bottles
Bottles Wash Bottles
Integrated safety vent cap eliminates dripping of vapours.
  • Caution note for each chemical
  • Coloured caps
  • Ergonomic spray angle
Bottles are manufactured from virgin LDPE resin, and come supplied with a polypropylene cap moulded with an ergonomic spray angle. The coloured caps allow for easy identification. A fill line is provided for user reference.
Please read the caution note mentioned over each bottle as per guideline.