VWR® Reagent Bottles, Wide Neck
Supplier: VWR
VWR®
VWRI215-4379EA 111 EUR
VWRI215-4379 VWRI215-4653 215-4689 215-4688 215-4402 215-4687 215-4401 215-4686 215-4400 215-4685 215-4684 VWRI215-4402 VWRI215-4400 215-4379 215-4653 215-4378 215-4693 215-4692 215-4691 215-4690 215-4381 215-4380 VWRI215-4399 VWRI215-4398 VWRI215-4397 VWRI215-4396 VWRI215-4395 VWRI215-4394 VWRI215-4393 VWRI215-4392 215-4389 215-4388 VWRI215-4391 215-4387 VWRI215-4390 215-4386 215-4385 215-4384 215-4383 215-4382 215-4392 215-4391 215-4390 VWRI215-4388 VWRI215-4387 VWRI215-4386 VWRI215-4385 VWRI215-4384 VWRI215-4383 VWRI215-4382 VWRI215-4381 VWRI215-4380 215-4399 215-4398 215-4397 215-4396 215-4395 215-4394 215-4393
VWR® Reagent Bottles, Wide Neck
Bottles Glass Bottles
Made from clear or amber soda-lime glass. Ideal for sampling, storage of powders or liquids, or other applications that do not involve high thermal stress.
  • Good chemical and physical properties
  • Clear or amber glass, delivered with black PP screw cap
  • Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
Certifications: Thread in accordance with DIN 168.